Assay Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271356.1 | NP_001258285.1 | 9 - 10 | 1654 | 91 | |
| NM_001271357.1 | NP_001258286.1 | 9 - 10 | 1397 | 91 | ||
| NM_146018.2 | NP_666130.1 | 9 - 10 | 1694 | 91 | ||
| GenBank mRNA | AK080933.1 | - | 9 - 10 | 1479 | 91 | |
| AK136071.1 | - | 9 - 10 | 1385 | 91 | ||
| AK148216.1 | - | 9 - 10 | 1448 | 91 | ||
| AK154040.1 | - | 9 - 10 | 1676 | 91 | ||
| AK172245.1 | - | 9 - 10 | 1452 | 91 | ||
| AK213109.1 | - | 2 - 3 | 145 | 91 | ||
| BC025820.1 | - | 9 - 10 | 1428 | 5099493 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Flcn
Genomic Map