Assay Details
Gene Symbol: Gm1647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC053975.1 | - | 1 - 2 | 196 | 133 | |
BC062821.1 | - | 1 - 2 | 187 | 133 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | karyopherin (importin) alpha 4 | Kpna4 |
1110058D08Rik, IPOA3 |
-4.1kb | Anti-sense | Chr. 3 - 69072221 - 69127092 on NCBI Build GRCm38 |
Downstream | ADP-ribosylation factor-like 14 | Arl14 |
9130014L17Rik, Arf7 |
71.7kb | Sense | Chr. 3 - 69222419 - 69223618 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
