Assay Details
| Gene Symbol: Crygc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007775.2 | NP_031801.1 | 2 - 3 | 282 | 90 | |
| GenBank mRNA | BC056454.1 | - | 1 - 2 | 261 | 6824225 | 90 |
| CJ066821.1 | - | 2 - 3 | 283 | 90 | ||
| EF426301.1 | - | 2 - 3 | 271 | 90 | ||
| M64544.1 | - | 1 - 2 | 234 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crygc
Genomic Map