Assay Details
| Gene Symbol: H2-DMa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010386.4 | NP_034516.3 | 5 - 6 | 1378 | 94 | |
| GenBank mRNA | AK146950.1 | - | 5 - 6 | 1379 | 94 | |
| AK160460.1 | - | 4 - 5 | 857 | 94 | ||
| BC049992.1 | - | 4 - 5 | 791 | 6394795 | 94 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
peptide antigen assembly with MHC class II protein complex
antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
positive regulation of germinal center formation
positive regulation of humoral immune response
protein complex assembly
immune response
protein transport
immunoglobulin mediated immune response
antigen processing and presentation
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive thymic T cell selection
positive regulation of T cell differentiation
inner ear development
positive regulation of immune response
chaperone mediated protein folding requiring cofactor
protein binding
MHC class II protein complex binding
immune system process
peptide antigen assembly with MHC class II protein complex
antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
positive regulation of germinal center formation
positive regulation of humoral immune response
protein complex assembly
immune response
protein transport
immunoglobulin mediated immune response
antigen processing and presentation
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive thymic T cell selection
positive regulation of T cell differentiation
inner ear development
positive regulation of immune response
chaperone mediated protein folding requiring cofactor
protein binding
MHC class II protein complex binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30414 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90794.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90792.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90792.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31152316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08764661 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08764651 | - | - |
| Whole Mouse Genome G4122A | A_51_P473734 | X90795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P473734 | X90794.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13316654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13316652 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13316651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31152359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13456815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13099324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13456794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13407412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13518624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13099327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31152316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444234 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459872_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422527_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444229 | - | - |
| Mouse Genome 430 2.0 Array | 1422527_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459872_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1459872_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422527_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14998_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14998_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700005837.mm.1 | NM_010386.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005837.mm.1 | TC1700000618.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422527_PM_at | - | - |
| Mouse Expression Set 430 | 1422527_at | - | - |
| Mouse Expression Set 430 | 1459872_x_at | - | - |
| Mouse Expression Set 430 | 1459872_PM_x_at | - | - |
| Murine Genome U74v2 Set | 93092_PM_at | - | - |
| Murine Genome U74v2 Set | 93092_at | - | - |
| Murine Genome U74v2 Set | 162346_f_at | - | - |
| Murine Genome U74v2 Set | 162346_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422527_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422527_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422527_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422527_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459872_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459872_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6290463 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290463 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777769 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777769 | - | - |
| mouse-6 v1.1 (Illumina) | scl014998.4_34-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290463 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014998.4_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290463 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2866856 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777769 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2866856 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014998.4_34-S | - | - |

H2-DMa
Genomic Map