Assay Details
| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195075.1 | NP_001182004.1 | 10 - 11 | 1061 | 74 | |
| GenBank mRNA | CD803575.1 | - | 1 - 2 | 116 | 74 | |
| CK780311.1 | - | 4 - 5 | 475 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl3
Genomic Map