Assay Details
| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195075.1 | NP_001182004.1 | 13 - 14 | 1474 | 94 | |
| GenBank mRNA | CD803575.1 | - | 4 - 5 | 529 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl3
Genomic Map