Assay Details
| Gene Symbol: Klhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195075.1 | NP_001182004.1 | 8 - 9 | 796 | 93 | |
| GenBank mRNA | CK780311.1 | - | 2 - 3 | 210 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl3
Genomic Map