Assay Details
| Gene Symbol: 5830454E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073359.1 | 1 | 326 | 80 | ||
| GenBank mRNA | AK018010.1 | - | 1 | 295 | 80 | |
| BC100520.1 | - | 1 | 311 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L14 | Rpl14 |
3100001N19Rik, AA407502, AL022816, CAG-ISL-7, CTG-B33, L14 |
-2.7kb | Sense | Chr. 9 - 120571517 - 120574653 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 39458 |
Gm39458 | 97.4kb | Sense | Chr. 9 - 120675443 - 120682465 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830454E08Rik
Genomic Map