Assay Details
| Gene Symbol: Gm8363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073378.1 | 1 | 118 | 103 | ||
| GenBank mRNA | EL606001.1 | - | 1 | 118 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 242 | Tmem242 |
1110008A10Rik, 2310046K16Rik, 5730437N04Rik |
-40.7kb | Sense | Chr. 17 - 5410864 - 5440260 on NCBI Build GRCm38 |
| Downstream |
zinc finger, DHHC domain containing 14 |
Zdhhc14 |
B530001K09Rik, New1cp |
8.9kb | Anti-sense | Chr. 17 - 5491946 - 5753891 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm8363
Genomic Map