Assay Details
| Gene Symbol: Kcnj15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073406.1 | 4 - 5 | 703 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31641 |
Gm31641 | -23.9kb | Anti-sense | Chr. 16 - 95184537 - 95263455 on NCBI Build GRCm38 | |
| Downstream | avian erythroblastosis virus E-26 (v-ets) oncogene related | Erg | D030036I24Rik | 67.3kb | Anti-sense | Chr. 16 - 95359169 - 95586593 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnj15
Genomic Map