| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200026938.mm.1 |
NM_001271724.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026938.mm.1 |
NM_013677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026938.mm.1 |
NM_013677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026938.mm.1 |
NR_073420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026938.mm.1 |
TC0200003204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200026943.mm.1 |
NM_001271724.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026943.mm.1 |
NM_013677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026943.mm.1 |
NM_013677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026943.mm.1 |
NR_073420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026943.mm.1 |
TC0200003204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200026944.mm.1 |
NM_001271724.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026944.mm.1 |
NM_013677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026944.mm.1 |
NM_013677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026944.mm.1 |
NR_073420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026944.mm.1 |
TC0200003204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200026937.mm.1 |
NM_001271724.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026937.mm.1 |
NM_013677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026937.mm.1 |
NM_013677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026937.mm.1 |
NR_073420.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200026937.mm.1 |
TC0200003204.mm.1 |
MTA TC |