Assay Details
| Gene Symbol: Aqp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073483.1 | 5 - 6 | 848 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 32017 |
Gm32017 | -163.4kb | Anti-sense | Chr. 9 - 70914146 - 70959418 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 32511 |
Gm32511 | 41.9kb | Anti-sense | Chr. 9 - 71168776 - 71208122 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Aqp9
Genomic Map