Assay Details
| Gene Symbol: Sema6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073549.1 | 5 - 6 | 739 | 82 | ||
| NR_073550.1 | 5 - 6 | 753 | 82 | |||
| NR_073551.1 | 6 - 7 | 802 | 82 | |||
| GenBank mRNA | AK031131.1 | - | 5 - 6 | 651 | 82 | |
| AK165085.1 | - | 5 - 6 | 666 | 82 | ||
| BC051979.1 | - | 6 - 7 | 708 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tumor necrosis factor, alpha-induced protein 8-like 2 |
Tnfaip8l2 |
1810019A08Rik, AW610835, TIPE2 |
-25.2kb | Anti-sense | Chr. 3 - 95139521 - 95142448 on NCBI Build GRCm38 |
| Downstream |
GA repeat binding protein, beta 2 |
Gabpb2 |
1810015F01Rik, 5830427M07, 9430006E19Rik, A430024B14Rik, AV050852, Gabpb2-1 |
13.7kb | Anti-sense | Chr. 3 - 95181766 - 95217951 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Sema6c
Genomic Map