Assay Details
| Gene Symbol: Aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_074087.1 | 3 - 4 | 385 | 126 | ||
| GenBank mRNA | BE685863.1 | - | 3 - 4 | 177 | 126 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
guanine nucleotide binding protein, alpha 11 |
Gna11 |
Dsk7, E430025L19 |
-19.0kb | Anti-sense | Chr. 10 - 81528732 - 81545046 on NCBI Build GRCm38 |
| Downstream |
transducin-like enhancer of split 2, homolog of Drosophila E(spl) |
Tle2 |
Grg2, mKIAA4188 |
10.0kb | Sense | Chr. 10 - 81574739 - 81592713 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Aes
Genomic Map