Assay Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_075088.1 | 4 - 5 | 685 | 103 | ||
| GenBank mRNA | AK160776.1 | - | 4 - 5 | 320 | 103 | |
| BC028655.1 | - | 4 - 5 | 685 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peptidylprolyl isomerase domain and WD repeat containing 1 | Ppwd1 |
4632422M10Rik, A330090G21Rik |
-5.3kb | Anti-sense | Chr. 13 - 104204869 - 104228844 on NCBI Build GRCm38 |
| Downstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 6 |
Adamts6 |
5031426K13, A930019D11Rik, ADAM-TS6, b2b1879.1Clo, b2b2029Clo, b2b2182Clo, b2b2187.1Clo, b2b2228Clo |
51.1kb | Sense | Chr. 13 - 104287347 - 104496694 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Cenpk
Genomic Map