Assay Details
| Gene Symbol: C030013G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077216.1 | 1 - 2 | 175 | 114 | ||
| GenBank mRNA | AA874255.1 | - | 1 - 2 | 99 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 22 (organic cation transporter), member 3 |
Slc22a3 |
EMT, Oct3, Orct3, Slca22a3 |
Anti-sense | Chr. 17 - 12419972 - 12507704 on NCBI Build GRCm38 | |
| Downstream |
solute carrier family 22 (organic cation transporter), member 2 |
Slc22a2 |
Oct2, Orct2 |
108.6kb | Sense | Chr. 17 - 12583958 - 12628488 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C030013G03Rik
Genomic Map