Assay Details
| Gene Symbol: Gm6297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077221.1 | 2 - 3 | 223 | 171 | ||
| NR_077226.1 | 2 - 3 | 385 | 171 | |||
| GenBank mRNA | AK139295.1 | - | 1 - 2 | 909 | 171 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933428C19 gene | 4933428C19Rik | -194.6kb | Sense | Chr. 4 - 40482840 - 40525548 on NCBI Build GRCm38 | |
| Downstream |
DnaJ (Hsp40) homolog, subfamily A, member 1 |
Dnaja1 |
Hsj2, Nedd7 |
801.0 | Anti-sense | Chr. 4 - 40722468 - 40737153 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6297
Genomic Map