Assay Details
| Gene Symbol: A230009B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077238.1 | 1 - 2 | 128 | 101 | ||
| GenBank mRNA | AK038759.1 | - | 1 - 2 | 132 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700110C19 gene | 1700110C19Rik | -126.9kb | Sense | Chr. 17 - 10324602 - 10329312 on NCBI Build GRCm38 | |
| Downstream | predicted gene 16169 | Gm16169 | OTTMUSG00000031251 | 333.6kb | Sense | Chr. 17 - 10807716 - 10826625 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A230009B12Rik
Genomic Map