Assay Details
| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102287.1 | 2 - 3 | 234 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cilia and flagella associated protein 74 | Cfap74 |
2010015L04Rik, A530045M11 |
Anti-sense | Chr. 4 - 155398208 - 155466823 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 52 | Tmem52 | 2310043I08Rik | 54.2kb | Anti-sense | Chr. 4 - 155469114 - 155470858 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

5830444B04Rik
Genomic Map