Assay Details
| Gene Symbol: C430002N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102293.1 | 1 - 2 | 116 | 93 | ||
| NR_102296.1 | 1 - 2 | 116 | 93 | |||
| GenBank mRNA | AA030139.1 | - | 1 - 2 | 77 | 93 | |
| AK049386.1 | - | 1 - 2 | 104 | 93 | ||
| AK089981.1 | - | 106 | 93 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E030011O05 gene | E030011O05Rik | -12.7kb | Anti-sense | Chr. 9 - 96731575 - 96752831 on NCBI Build GRCm38 | |
| Downstream | 2-phosphoxylose phosphatase 1 | Pxylp1 |
9430094M07Rik, Acpl2, BB177120, C130099A20Rik |
53.6kb | Anti-sense | Chr. 9 - 96823336 - 96892680 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C430002N11Rik
Genomic Map