Assay Details
| Gene Symbol: A330033J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102303.1 | 1 - 2 | 703 | 119 | ||
| GenBank mRNA | AK034282.1 | - | 1 - 2 | 703 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36346 |
Gm36346 | Sense | Chr. 13 - 48107897 - 48111641 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA A330048O09 gene | A330048O09Rik | 9.8kb | Sense | Chr. 13 - 48272418 - 48273884 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A330033J07Rik
Genomic Map