Assay Details
| Gene Symbol: Hrh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102309.1 | 1 - 2 | 509 | 70 | ||
| GenBank mRNA | BC145125.1 | - | 1 - 2 | 276 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosome associated GTPase 2 | Mtg2 |
1810011P19Rik, 2900056P18Rik, D2Bwg0647e, Gtpbp5 |
-16.8kb | Anti-sense | Chr. 2 - 180070593 - 180085902 on NCBI Build GRCm38 |
| Downstream | oxysterol binding protein-like 2 | Osbpl2 |
C130070J12Rik, ORP-2, Orp2, mKIAA0772 |
15.0kb | Anti-sense | Chr. 2 - 180119366 - 180162680 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Hrh3
Genomic Map