Assay Details
| Gene Symbol: D630024D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102310.1 | 3 - 4 | 559 | 91 | ||
| NR_102311.1 | 3 - 4 | 514 | 91 | |||
| GenBank mRNA | AK158233.1 | - | 3 - 4 | 497 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | biorientation of chromosomes in cell division 1 | Bod1 |
2310022M17Rik, BOD2.1, Fam44b |
-146.1kb | Sense | Chr. 11 - 31665150 - 31671862 on NCBI Build GRCm38 |
| Downstream | cytoplasmic polyadenylation element binding protein 4 | Cpeb4 |
Cpe-bp4, Cpeb-4, mCPEB-4 |
52.3kb | Anti-sense | Chr. 11 - 31870884 - 31935635 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

D630024D03Rik
Genomic Map