Assay Details
| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102319.1 | 2 - 3 | 725 | 175 | ||
| GenBank mRNA | AK145081.1 | - | 2262 | 175 | ||
| FJ744746.1 | - | 2 - 3 | 647 | 175 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35909 |
Gm35909 | -69.2kb | Sense | Chr. 14 - 105518096 - 105523485 on NCBI Build GRCm38 | |
| Downstream | microRNA 6390 | Mir6390 |
Gm27562, mmu-mir-6390 |
29.7kb | Anti-sense | Chr. 14 - 105623642 - 105623770 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9330188P03Rik
Genomic Map