Assay Details
| Gene Symbol: 6430710C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102348.1 | 2 - 3 | 365 | 120 | ||
| NR_102349.1 | 3 - 4 | 219 | ||||
| GenBank mRNA | AK019889.1 | - | 3 - 4 | 219 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 8430437L04 gene | 8430437L04Rik | -58.7kb | Anti-sense | Chr. 2 - 72703510 - 72729468 on NCBI Build GRCm38 | |
| Downstream | trans-acting transcription factor 3 | Sp3 | D130027J01Rik | 136.2kb | Sense | Chr. 2 - 72936430 - 72980446 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

6430710C18Rik
Genomic Map