Assay Details
| Gene Symbol: 6430710C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102348.1 | 4 - 5 | 539 | 83 | ||
| GenBank mRNA | AK018302.1 | - | 4 - 5 | 495 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 8430437L04 gene | 8430437L04Rik | -18.1kb | Anti-sense | Chr. 2 - 72703510 - 72729468 on NCBI Build GRCm38 | |
| Downstream | trans-acting transcription factor 3 | Sp3 | D130027J01Rik | 149.0kb | Sense | Chr. 2 - 72936430 - 72980446 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

6430710C18Rik
Genomic Map