Assay Details
| Gene Symbol: AU015791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102381.1 | 4 - 5 | 476 | 111 | ||
| GenBank mRNA | AU017690.1 | - | 1 - 2 | 111 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D430019H16 gene | D430019H16Rik | AI852661 | -21.2kb | Sense | Chr. 12 - 105453856 - 105493095 on NCBI Build GRCm38 |
| Downstream |
bradykinin receptor, beta 2 |
Bdkrb2 |
B(2), B2, B2R, BK-2, BK2, BK2R, BKR2, BRB2 |
47.9kb | Sense | Chr. 12 - 105563172 - 105595244 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AU015791
Genomic Map