Assay Details
| Gene Symbol: Fgf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102395.1 | 2 - 3 | 247 | 89 | ||
| GenBank mRNA | AK078245.1 | - | 2 - 3 | 66 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small integral membrane protein 23 | Smim23 | 1700008A04Rik | -309.6kb | Sense | Chr. 11 - 32820376 - 32824594 on NCBI Build GRCm38 |
| Downstream | nucleophosmin 1 | Npm1 |
B23, NO38, Npm |
5.4kb | Sense | Chr. 11 - 33152498 - 33163206 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fgf18
Genomic Map