Assay Details
| Gene Symbol: AA413626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102683.1 | 1 | 116 | 101 | ||
| GenBank mRNA | AA413626.1 | - | 1 | 116 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1 (C. elegans) | Nipsnap1 | -24.3kb | Sense | Chr. 11 - 4873965 - 4894202 on NCBI Build GRCm38 | |
| Downstream |
neurofilament, heavy polypeptide |
Nefh |
NF-H, NF200, Nfh, mKIAA0845 |
19.8kb | Anti-sense | Chr. 11 - 4938754 - 4948064 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AA413626
Genomic Map