Assay Details
| Gene Symbol: 5430425K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103550.1 | 1 - 2 | 250 | 79 | ||
| GenBank mRNA | DV053914.1 | - | 1 - 2 | 268 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | arrestin domain containing 3 | Arrdc3 |
AI450344, mKIAA1376 |
-44.4kb | Sense | Chr. 13 - 80883338 - 80896042 on NCBI Build GRCm38 |
| Downstream | predicted gene 4211 | Gm4211 | 16.4kb | Anti-sense | Chr. 13 - 80957664 - 80961434 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

5430425K12Rik
Genomic Map