Assay Details
| Gene Symbol: Gnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104298.1 | 5 - 6 | 608 | 88 | ||
| GenBank mRNA | BC018560.1 | - | 5 - 6 | 495 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 19 |
Snord19 | MBII-108 | -50.0 | Sense | Chr. 14 - 31016219 - 31016272 on NCBI Build GRCm38 |
| Downstream | polybromo 1 | Pbrm1 |
2610016F04Rik, AI507524, BAF180, Pb1 |
2.2kb | Anti-sense | Chr. 14 - 31019138 - 31121592 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gnl3
Genomic Map