Assay Details
| Gene Symbol: Gnb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081682.2 | NP_001075151.1 | 4 - 5 | 499 | 116 | |
| NM_001285494.1 | NP_001272423.1 | 4 - 5 | 441 | 116 | ||
| NM_023120.5 | NP_075609.2 | 4 - 5 | 441 | 116 | ||
| NR_104347.1 | 4 - 5 | 441 | 116 | |||
| GenBank mRNA | AF301595.1 | - | 4 - 5 | 392 | 116 | |
| AK166195.1 | - | 4 - 5 | 425 | 116 | ||
| AK171253.1 | - | 4 - 5 | 421 | 116 | ||
| AK173227.1 | - | 4 - 5 | 424 | 116 | ||
| BC024635.1 | - | 4 - 5 | 410 | 4193427 | 116 | |
| BC037676.1 | - | 3 - 4 | 327 | 3493788 | 116 | |
More Information
Set Membership: |
|

Gnb1l
Genomic Map