Assay Details
| Gene Symbol: Gm28042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104353.1 | 5 - 6 | 650 | 90 | ||
| GenBank mRNA | AB195274.1 | - | 5 - 6 | 640 | 90 | |
| AB195275.1 | - | 5 - 6 | 640 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase binding protein 1 | Mapkbp1 |
2810483F24Rik, AW123212, JNKBP-1, Jnkbp1, mKIAA0596 |
-3.9kb | Sense | Chr. 2 - 119972388 - 120027403 on NCBI Build GRCm38 |
| Downstream |
phospholipase A2, group IVB (cytosolic) |
Pla2g4b |
A030011C02Rik, Gm1013, cPLA2-beta |
1.8kb | Sense | Chr. 2 - 120033433 - 120043033 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB195274.1, AB195275.1 transcripts.
Set Membership: |
|

Gm28042
Genomic Map