Assay Details
| Gene Symbol: Chchd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104356.1 | 2 - 3 | 170 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pleiomorphic adenoma gene 1 | Plag1 | -2.8kb | Anti-sense | Chr. 4 - 3898769 - 3938434 on NCBI Build GRCm38 | |
| Downstream |
short chain dehydrogenase/reductase family 16C, member 5 |
Sdr16c5 |
Rdhe2, Scdr9 |
53.1kb | Anti-sense | Chr. 4 - 3995942 - 4019663 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Chchd7
Genomic Map