Assay Details
| Gene Symbol: Exosc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104358.1 | 2 - 3 | 346 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
branched chain ketoacid dehydrogenase E1, alpha polypeptide |
Bckdha | -4.4kb | Anti-sense | Chr. 7 - 25629852 - 25658784 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 91 | Tmem91 |
A830041P22Rik, DSPC3 |
4.7kb | Anti-sense | Chr. 7 - 25669139 - 25679815 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Exosc5
Genomic Map