Assay Details
| Gene Symbol: Osgepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104360.1 | 2 - 3 | 474 | 65 | ||
| GenBank mRNA | BC030671.1 | - | 2 - 3 | 444 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ORM1-like 1 (S. cerevisiae) | Ormdl1 | C730042F17Rik | -4.6kb | Sense | Chr. 1 - 53297095 - 53310245 on NCBI Build GRCm38 |
| Downstream | asparagine synthetase domain containing 1 | Asnsd1 | 2210409M21Rik | 26.3kb | Anti-sense | Chr. 1 - 53344617 - 53352752 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Osgepl1
Genomic Map