Assay Details
| Gene Symbol: 1700024P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104376.1 | 16 - 17 | 2076 | 119 | ||
| GenBank mRNA | BC144858.1 | - | 16 - 17 | 2077 | 9053406 | 120 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
complement component 8, alpha polypeptide |
C8a | -128.8kb | Anti-sense | Chr. 4 - 104815679 - 104876486 on NCBI Build GRCm38 | |
| Downstream |
protein kinase, AMP-activated, alpha 2 catalytic subunit |
Prkaa2 |
2310008I11Rik, A830082D05, AMPKalpha2 |
19.1kb | Anti-sense | Chr. 4 - 105029650 - 105109898 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700024P16Rik
Genomic Map