Assay Details
| Gene Symbol: Plekhg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104381.1 | 1 - 2 | 90 | 74 | ||
| GenBank mRNA | AK154912.1 | - | 1 - 2 | 82 | 74 | |
| BY151802.1 | - | 1 - 2 | 88 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleolar protein 9 | Nol9 |
4632412I24Rik, 6030462G04Rik, AI449622, AW490720 |
-25.4kb | Sense | Chr. 4 - 152039274 - 152061494 on NCBI Build GRCm38 |
| Downstream |
tumor necrosis factor receptor superfamily, member 25 |
Tnfrsf25 |
APO-3, DDR3, DR3, LARD, TR3, TRAMP, Tnfrsf12, WSL-1, WSL-LR, Wsl |
27.8kb | Sense | Chr. 4 - 152115528 - 152120119 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhg5
Genomic Map