Assay Details
| Gene Symbol: Eif4e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104407.1 | 1 - 2 | 99 | 78 | ||
| GenBank mRNA | DQ279476.1 | - | 1 - 2 | 96 | 78 | |
| DQ279477.1 | - | 1 - 2 | 96 | 78 | ||
| DQ279479.1 | - | 1 - 2 | 94 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 16249 | Gm16249 | OTTMUSG00000032672 | -907.0 | Anti-sense | Chr. 13 - 54777514 - 54783091 on NCBI Build GRCm38 |
| Downstream | tetraspanin 17 | Tspan17 |
2210021G21Rik, AI047581, Fbxo23, Tm4sf17 |
5.1kb | Sense | Chr. 13 - 54789352 - 54796896 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Eif4e1b
Genomic Map