Assay Details
| Gene Symbol: Fuom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104415.1 | 5 - 6 | 527 | 86 | ||
| GenBank mRNA | AK044731.1 | - | 5 - 6 | 492 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline-rich acidic protein 1 | Prap1 | Upa | -612.0 | Anti-sense | Chr. 7 - 140093396 - 140097203 on NCBI Build GRCm38 |
| Downstream |
enoyl Coenzyme A hydratase, short chain, 1, mitochondrial |
Echs1 | C80529 | 5.8kb | Sense | Chr. 7 - 140105723 - 140116423 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fuom
Genomic Map