Assay Details
| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104441.1 | 9 - 10 | 1218 | 77 | ||
| GenBank mRNA | AK138661.1 | - | 9 - 10 | 1217 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hydrogen voltage-gated channel 1 | Hvcn1 |
0610039P13Rik, AI450555, BTS, HV1, Vsop., mVSOP |
-4.2kb | Anti-sense | Chr. 5 - 122206807 - 122242297 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700112N08 gene | 1700112N08Rik | 27.3kb | Anti-sense | Chr. 5 - 122275323 - 122279719 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tctn1
Genomic Map