Assay Details
| Gene Symbol: Cuta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104458.1 | 4 - 5 | 495 | 102 | ||
| GenBank mRNA | AK007580.1 | - | 3 - 4 | 538 | 102 | |
| AK007912.1 | - | 4 - 5 | 484 | 102 | ||
| AK145185.1 | - | 563 | 102 | |||
| AK207668.1 | - | 1 - 2 | 143 | 102 | ||
| BC028531.1 | - | 4 - 5 | 500 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PHD finger protein 1 | Phf1 |
AW557215, D17Ertd455e, Pcl1, Phf2, Tctex3 |
-510.0 | Anti-sense | Chr. 17 - 26933127 - 26937908 on NCBI Build GRCm38 |
| Downstream | synaptic Ras GTPase activating protein 1 homolog (rat) | Syngap1 |
Gm1963, Syngap |
2.8kb | Anti-sense | Chr. 17 - 26941408 - 26972434 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Cuta
Genomic Map