Assay Details
| Gene Symbol: Banf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104482.1 | 1 - 2 | 313 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cystatin E/M | Cst6 |
1110017E11Rik, N28197, ichq |
-15.1kb | Sense | Chr. 19 - 5344705 - 5349574 on NCBI Build GRCm38 |
| Downstream | squamous cell carcinoma antigen recognized by T cells 1 | Sart1 | U5-110K | 10.6kb | Sense | Chr. 19 - 5377523 - 5389420 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Banf1
Genomic Map