Assay Details
| Gene Symbol: Mir6343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105761.1 | 1 | 38 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 17751 |
Gm17751 | -7.5kb | Sense | Chr. 1 - 76489133 - 76502028 on NCBI Build GRCm38 | |
| Downstream | Eph receptor A4 | Epha4 |
2900005C20Rik, AI385584, Cek8, Hek8, Sek, Sek1, Tyro1, rb |
857.5kb | Anti-sense | Chr. 1 - 77367182 - 77515103 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6343
Genomic Map