Assay Details
| Gene Symbol: Mir6351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105769.1 | 1 | 48 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 6030407O03 gene | 6030407O03Rik | Sense | Chr. 1 - 73618381 - 73864466 on NCBI Build GRCm38 | ||
| Downstream | tensin 1 | Tns1 |
1110018I21Rik, 1200014E20Rik, AI648117, E030018G17Rik, E030037J05Rik, Tns |
54.7kb | Sense | Chr. 1 - 73910231 - 74124535 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6351
Genomic Map