Assay Details
| Gene Symbol: Mir6352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105770.1 | 1 | 59 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6343 | Mir6343 |
Gm27304, mmu-mir-6343 |
-987.0kb | Anti-sense | Chr. 1 - 76509560 - 76509643 on NCBI Build GRCm38 |
| Downstream | paired box 3 | Pax3 |
Pax-3, Sp, splotch |
604.5kb | Sense | Chr. 1 - 78101267 - 78197547 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6352
Genomic Map