Assay Details
| Gene Symbol: Mir6358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105776.1 | 1 | 50 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 7C | Zbtb7c |
B230208J24Rik, Kr-pok, Zbtb36 |
-22.0kb | Sense | Chr. 18 - 75820178 - 76148564 on NCBI Build GRCm38 |
| Downstream | SMAD family member 2 | Smad2 |
7120426M23Rik, Madh2, Madr2, Smad-2, mMad2 |
70.6kb | Sense | Chr. 18 - 76241251 - 76311748 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6358
Genomic Map