Assay Details
| Gene Symbol: Mir6369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105788.1 | 1 | 59 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26555 |
Gm26555 | -1.8kb | Anti-sense | Chr. 13 - 58279261 - 58311198 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2210016F16 gene | 2210016F16Rik | 66.9kb | Sense | Chr. 13 - 58380046 - 58385225 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mir6369
Genomic Map