Assay Details
| Gene Symbol: Mir6387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105809.1 | 1 | 54 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tribbles homolog 2 (Drosophila) | Trib2 |
AW319517, TRB-2, TRB2 |
Sense | Chr. 12 - 15791725 - 15816962 on NCBI Build GRCm38 | |
| Downstream | lipin 1 | Lpin1 |
4631420P06, Kiaa0188, Lipin1, fld, mKIAA0188 |
734.6kb | Sense | Chr. 12 - 16535669 - 16646968 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6387
Genomic Map