Assay Details
| Gene Symbol: Mir6389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105812.1 | 1 | 55 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
gamma-aminobutyric acid (GABA) A receptor, subunit alpha 5 |
Gabra5 | A230018I05Rik | -71.0kb | Anti-sense | Chr. 7 - 57407669 - 57510100 on NCBI Build GRCm38 |
| Downstream |
gamma-aminobutyric acid (GABA) A receptor, subunit beta 3 |
Gabrb3 |
A230092K12Rik, AW049585, Cp1, Gabrb-3, beta3 |
9.4kb | Sense | Chr. 7 - 57590518 - 57828802 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6389
Genomic Map